Learn More
Sino Biological Recombinant Human FGFR2 Protein (His & hFc Tag), HPLC-verified
Supplier:  Sino Biological 10824H03H100
Encompass_Preferred
A DNA sequence encoding the human FGFR2 (NP_000132.3) extracellular domain (Met 1-Glu 377) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5672