Learn More
Sino Biological Recombinant Human FGFR2 Protein (aa 400-821, His & GST Tag)
Supplier:  Sino Biological 10824H20B150
Encompass_Preferred
A DNA sequence encoding the human FGFR2 (NP_000132.3) cytoplasmic domain (Met 400-Thr 821) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-5677