Learn More
Sino Biological Recombinant Human RSK3 Protein (GST Tag)
Supplier:  Sino Biological 10833H09B50
Encompass_Preferred
A DNA sequence encoding the human RPS6KA2 isoform 1 (Q15349-1) (Met 1-Leu 733) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-5687