Learn More
Sino Biological Recombinant Human SLAM/CD150 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10837H08H100
Encompass_Preferred
A DNA sequence encoding the human SLAM (NP_003028.1) precursor (Met 1-Pro 258) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5691