Learn More
Sino Biological Recombinant Human 14-3-3 sigma Protein
Supplier:  Sino Biological 10838HNCE100
Encompass_Preferred
A DNA sequence encoding the human SFN (NP_006133.1) (Met 1-Ser 248) was expressed, with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-5695