Learn More
Sino Biological Recombinant Human DKKL1 Protein (hFc Tag)
Supplier:  Sino Biological 10840H01H50
Encompass_Preferred
A DNA sequence encoding the secreted form of human DKKL1 (Q9UK85) (Ala 31-Leu 242) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-5700