Learn More
Sino Biological Recombinant Human 14-3-3 epsilon Protein (GST Tag)
Supplier:  Sino Biological 10842H09E100
Encompass_Preferred
A DNA sequence encoding the human YWHAE (NP_006752.1) (Met 1-Gln 255) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-5703