Learn More
Sino Biological Recombinant Human 14-3-3 epsilon Protein
Supplier:  Sino Biological 10842HNCE100
Encompass_Preferred
A DNA sequence encoding the human YWHAE (NP_006752.1) (Met 1-Gln 255) was expressed, with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-5705