Learn More
Sino Biological Recombinant Human 14-3-3 beta Protein (GST Tag)
Supplier:  Sino Biological 10843H09E100
Encompass_Preferred
A DNA sequence encoding the human YWHAB (NP_003395.1) (Met 1-Asn 246) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-5707