Learn More
Sino Biological Recombinant Human Nectin 3 Protein (His Tag)
Supplier:  Sino Biological 10852H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human PVRL3 (NP_056295.1) (Met 1-Asp 400) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5719