Learn More
Sino Biological Recombinant Human MFGE8 Protein (His Tag)
Supplier:  Sino Biological 10853H08B100
Encompass_Preferred
A DNA sequence encoding the human MFGE8 isoform 1 (Q08431-1) (Met 1-Cys 387) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5721