Learn More
Sino Biological Recombinant Human GPR114 Protein (hFc Tag)
Supplier:  Sino Biological 10854H02H100
Encompass_Preferred
A DNA sequence encoding the human GPR114 (Q8IZF4) (Met1-Gly184) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5724