Learn More
Sino Biological Recombinant Human HDAC8 Protein (GST Tag)
Supplier:  Sino Biological 10864H09B100
Encompass_Preferred
A DNA sequence encoding the full length of human HDAC8 isoform 1 (NP_060956.1) (Met 1-Val 377) was expressed, fused with the GST tag at the C-terminus.
Catalog No. 50-161-5726