Learn More
Sino Biological Recombinant Human TNFR1/CD120a Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10872H08H100
Encompass_Preferred
A DNA sequence encoding the human TNFRI (NP_001056.1) extracellular domain (Met 1-Thr 211) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5746