Learn More
Sino Biological Recombinant Human CXCL4/PF4 Protein
Supplier:  Sino Biological 10874HNAE100
Encompass_Preferred
A DNA sequence encoding the human CXCL4 (NP_002610.1) (Glu 32-Ser 101) was expressed.
Catalog No. 50-161-5750