Learn More
Sino Biological Recombinant Human CXCL1 Protein (His & SUMO Tag)
Supplier:  Sino Biological 10877H28E50
Encompass_Preferred
A DNA sequence encoding the mature form of human CXCL1 (P09341-1) (Ala 35-Asn 107) was fused with a N-terminal polyhistide-tagged SUMO tag at the N-terminus.
Catalog No. 50-161-5758