Learn More
Sino Biological Recombinant Human Ephrin B2 Protein
Supplier:  Sino Biological 10881HCCH100
Encompass_Preferred
A DNA sequence encoding the human EFNB2 (NP_004084.1) (Met1-Ala229) was expressed with six amino acids (ENLYFQ) at the C-terminus was expressed and purified.
Catalog No. 50-161-5772