Learn More
Sino Biological Recombinant Human MAP1D Protein (His Tag)
Supplier:  Sino Biological 10883H08B50
Encompass_Preferred
A DNA sequence encoding the human MAP1D (NP_954697.1) (Arg 44-Ala 335) was expressed, with an initial Met at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5780