Learn More
Sino Biological Recombinant Human CXCL9/MIG Protein
Catalog No. 501615790
Encompass_Preferred
A DNA sequence encoding the human CXCL9 (Q07325) (Thr 23-Thr 125) was expressed and purified.
Catalog No. 50-161-5790 Supplier Sino Biological Supplier No. 10888HNAE50
A DNA sequence encoding the human CXCL9 (Q07325) (Thr 23-Thr 125) was expressed and purified.