Learn More
Sino Biological Recombinant Human BAMBI Protein (His Tag)
Supplier:  Sino Biological 10890H08H50
Encompass_Preferred
A DNA sequence encoding the human BAMBI extracellular domain (NP_036474.1) (Met 1-Ala 152) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-5794