Learn More
Sino Biological Recombinant Human Ephrin-B1 Protein (His Tag)
Supplier:  Sino Biological 10894H08H100
Encompass_Preferred
A DNA sequence encoding the human EFNB1 (NP_004420.1) extracellular domain (Met 1-Lys 237) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5797