Learn More
Sino Biological Recombinant Human IL-17RA Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10895H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human IL17RA (NP_055154.3) (Met 1-Trp 320) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5803