Learn More
Sino Biological Recombinant Human ADAM12 Protein (His Tag)
Supplier:  Sino Biological 10896H08H20
Encompass_Preferred
A DNA sequence encoding the human ADAM12 isoform 1 (NP_003465.3) extracellular domain (Met 1-Ser 513) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5806