Learn More
Sino Biological Recombinant Human MIP-5/CCL15 Protein (aa 22-113, His Tag)
Supplier:  Sino Biological 10898H08E50
Encompass_Preferred
A DNA sequence encoding the mature form of human CCL15 (Q16663-1) (Gln 22-Ile 113) was expressed, with a polyhistide tag at the C-terminus.
Catalog No. 50-161-5812