Learn More
Sino Biological Recombinant Human CCL4/MIP-1 Protein (His Tag)
Supplier:  Sino Biological 10899H08Y20
Encompass_Preferred
A DNA sequence encoding human CCL4 (NP_002975.1) (Ala24-Gln92) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5814