Learn More
Sino Biological Recombinant Human CCL5/RANTES Protein (His & mucin Tag)
Supplier:  Sino Biological 10900H14H50
Encompass_Preferred
A DNA sequence encoding the human RANTES (NP_002976.2) (Met 1-Ser 91) was fused with a C-terminal human Fractalkine mucin-stalk (Phe 103-Gln 341) followed by a polyhistidine tag.
Catalog No. 50-161-5816