Learn More
Sino Biological Recombinant Human CCL5/RANTES Protein
Catalog No. 501615817
Encompass_Preferred
A DNA sequence encoding the human CCL5 (NP_002976.2) (Ser24-Ser91) was expressed with an initial Met.
Catalog No. 50-161-5817 Supplier Sino Biological Supplier No. 10900HNAE100
A DNA sequence encoding the human CCL5 (NP_002976.2) (Ser24-Ser91) was expressed with an initial Met.