Learn More
Sino Biological Recombinant Human CCL5/RANTES Protein
Catalog No. 501615818
Encompass_Preferred
A DNA sequence encoding the human CCL5 (NP_002976.2) (Ser24-Ser91) was expressed with an initial Met.
Catalog No. 50-161-5818 Supplier Sino Biological Supplier No. 10900HNAE20
Add to Cart
Add to Cart
A DNA sequence encoding the human CCL5 (NP_002976.2) (Ser24-Ser91) was expressed with an initial Met.