Learn More
Sino Biological Recombinant Human FGF-21 Protein (His Tag)
Supplier:  Sino Biological 10911H07E50
Encompass_Preferred
A DNA sequence encoding the mature form of human FGF21 (NP_061986.1) (His 29-Ser 209) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-5844