Learn More
Sino Biological Recombinant Human OSTM1 Protein (His Tag)
Supplier:  Sino Biological 10913H08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human OSTM1 (NP_054747.2) (Met 1-Pro 284) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-5848