Learn More
Sino Biological Recombinant Human TIGIT Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10917H02H50
Encompass_Preferred
A DNA sequence encoding the human TIGIT (NP_776160.2) (Met1-Pro141) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5852