Learn More
Sino Biological Recombinant Human uPAR/PLAUR Protein (His Tag)
Supplier:  Sino Biological 10925H08H100
Encompass_Preferred
A DNA sequence encoding the human UPAR isoform 1 (Q03405-1) (Met 1-Arg 303) without the pro peptide was expressed, with a carboxy-terminal polyhistidine tag.
Catalog No. 50-161-5862