Learn More
Sino Biological Recombinant Human SPARC Protein (His Tag), HPLC-verified
Catalog No. 501615866
Encompass_Preferred
A DNA sequence encoding the human SPARC (NP_003109.1) precursor (Met 1-Ile 303) with a carboxy-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5866 Supplier Sino Biological Supplier No. 10929H08H100
A DNA sequence encoding the human SPARC (NP_003109.1) precursor (Met 1-Ile 303) with a carboxy-terminal polyhistidine tag was expressed.