Learn More
Sino Biological Recombinant Human TIMP-1 Protein (hFc Tag)
Supplier:  Sino Biological 10934H01H100
Encompass_Preferred
A DNA sequence encoding the human TIMP1 (NP_003245.1) (Cys24-Ala207) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-5871