Learn More
Sino Biological Recombinant Human TIMP-1 Protein (His Tag)
Supplier:  Sino Biological 10934H08H20
Encompass_Preferred
A DNA sequence encoding the human TIMP1 (NP_003245.1) (Met 1-Ala 207) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5875