Learn More
Sino Biological Recombinant Human GDF-15 Protein (Mature Form, His Tag)
Supplier:  Sino Biological 10936H07Y100
Encompass_Preferred
A DNA sequence encoding the mature form of human GDF15 (NP_004855.2) (Ala197-Ile308) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-5880