Learn More
Sino Biological Recombinant Human S100A6 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10939H08B50
Encompass_Preferred
A DNA sequence encoding the human S100A6 (NP_055439.1) (Met 1-Gly 90) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-5885