Learn More
Sino Biological Recombinant Human C1QB Protein (His Tag)
Supplier:  Sino Biological 10941H08B100
Encompass_Preferred
A DNA sequence encoding the human C1QB (NP_000482.3) precursor (Met 1-Ala 253) was expressed, with a carboxy-terminal polyhistidine tag.
Catalog No. 50-161-5888