Learn More
Sino Biological Recombinant Human IL-13 R alpha 1/IL13RA1 Protein (His & hFc Tag)
Supplier:  Sino Biological 10943H03H100
Encompass_Preferred
A DNA sequence encoding the human IL13Rα1 (NP_001551.1) extracellular domain (Met 1-Thr 343) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5892