Learn More
Sino Biological Recombinant Human IL-10 Protein (His Tag)
Supplier:  Sino Biological 10947H07H100
Encompass_Preferred
A DNA sequence encoding the human IL10 (NP_000563.1) (Ser 19-Asn 178) was expressed, with a N-terminal polyhistidine tag.
Catalog No. 50-161-5906