Learn More
Sino Biological Recombinant Human IL-10 Protein, HPLC-verified
Supplier:  Sino Biological 10947HNAE1
Encompass_Preferred
A DNA sequence encoding human IL10 (NP_000563.1) (Ser19-Asn178) was expressed.
Catalog No. 50-161-5909