Learn More
Sino Biological Recombinant Human alpha-2-macroglobulin Protein (His Tag)
Supplier:  Sino Biological 10952H08B50
Encompass_Preferred
A DNA sequence encoding the human A2M (NP_000005.2) (Met 1-Ala 1474) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5919