Learn More
Sino Biological Recombinant Human Prostatic Acid Phosphatase/ACPP Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10959H08H20
Encompass_Preferred
A DNA sequence encoding the human ACPP (NP_001127666.1) (Met 1-Lys 382) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5927