Learn More
Sino Biological Recombinant Human Von Willebrand Factor/vWF Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10973H08C100
Encompass_Preferred
A DNA sequence encoding the pro form of human von Willebrand factor (AAB59458.1) (Met1-Lys2813) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5937