Learn More
Sino Biological Recombinant Human IL-7R alpha/CD127 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10975H08H100
Encompass_Preferred
A DNA sequence encoding the human IL7Rα IL7R (P16871-1) (Met1-Gly236, with mutations I66T, V138I) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5947