Learn More
Sino Biological Recombinant Human CD3 delta/CD3D Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10981H08H50
Encompass_Preferred
A DNA sequence encoding the human CD3D extracellular domain (NP_000723.1) (Met 1-Ala 105) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5962