Learn More
Sino Biological Recombinant Human NAMPT Protein (His & GST Tag), HPLC-verified
Supplier:  Sino Biological 10990H20B50
Encompass_Preferred
A DNA sequence encoding the human NAMPT (P43490) (Met 1-His 491) was fused with a N-terminal polyhistidine tag followed by a GST tag.
Catalog No. 50-161-5975