Learn More
Sino Biological Recombinant Human CRISP3 Protein (His Tag)
Supplier:  Sino Biological 10996H08H50
Encompass_Preferred
A DNA sequence encoding the human CRISP3 (P54108.1) (Met1-Tyr245) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5985