Learn More
Sino Biological Recombinant Human KIAA0101 Protein (His Tag)
Supplier:  Sino Biological 10997H07E50
Encompass_Preferred
A DNA sequence encoding the human PAF isoform 1 (NP_055551.1) (Met 1-Glu 111) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-5987