Learn More
Electron Microscopy Sciences CoolSafe™ Cooling Brick 1°C
SDP
Catalog No. 5033416
Encompass_Preferred
Each Cool Brick allows up to 10 hours of benchtop temperature stability. Each Cool Brick allows up to 10 hours of benchtop temperature stability.
Catalog No. 50-334-16 Supplier Electron Microscopy Sciences Supplier No. 6195230
Each Cool Brick allows up to 10 hours of benchtop temperature stability. Each Cool Brick allows up to 10 hours of benchtop temperature stability.